Capillary columns, CP-Sil
Proveedor: VARIAN
CP7751 CP7741 CP8688 CP7691 CP8014 CP8529 CP8730 CP7130 CP7441 CP7443 CP7444 CP7452 CP7454 CP7619 CP7620 CP7625 CP7627 CP7629 CP7635 CP7645 CP7646 CP7647 CP7649 CP7657 CP7659 CP7666 CP7670 CP7671 CP7675 CP7676 CP7685 CP7688 CP7693 CP7695 CP7700 CP7709 CP7710 CP7711 CP7712 CP7721 CP7722 CP7730 CP7731 CP7739 CP7742 CP7750 CP7758 CP7760 CP7762 CP7770 CP7829 CP7895 CP7906 CP7907 CP7917 CP7926 CP7936 CP7937 CP7946 CP7947 CP7977 CP8510 CP8521 CP8530 CP8540 CP8541 CP8560 CP8562 CP8673 CP8674 CP8677 CP8689 CP8690 CP8710 CP8712 CP8716 CP8722 CP8735 CP8736 CP8737 CP8741 CP8742 CP8743 CP8744 CP8751 CP8752 CP8753 CP8754 CP8756 CP8760 CP8761 CP8762 CP8770 CP8771 CP8772 CP8775 CP8780 CP8781 CP8791 CP8796 CP8799 CP8842 CP8870 CP8871 CP7681 CP7761 CP8431 CP7702 CP7680 CP7451 CP7453 CP7752 CP7135 CP7140 CP6666 CP7196 CP7630 CP7442 CP7443I5 CP7453I5 CP7732 CP7740 CP7749 CP7749I5 CP7750I5 CP7751I5 CP7741I5 CP7719 CP7720 CP7789 CP7809 CP7779 CP7697 CP7769 CP7770I5 CP7771 CP7772 CP7631 CP7621 CP7622 CP7636 CP7637 CP7608 CP8430 CP7680I5 CP7682 CP7684 CP8542 CP7690 CP7690I5 CP7691I5 CP7692 CP7694 CP7696 CP7640 CP7641 CP8511 CP8531 CP7839 CP7813 CP7896 CP7884 CP8690I5 CP8676 CP8678 CP8685 CP8687 CP8687I5 CP8688I5 CP8675 CP8735I5 CP8736I5 CP8741I5 CP8750 CP7656 CP7667 CP7670I5 CP7672 CP6680 CP6670 CP7150 CP6681 CP6671 CP7340 CP7160 CP7170 CP7310 CP7311 CP7685I5 CP8774
561-7751EA 1810 EUR
561-7751 561-7741 561-8688 VARICP7691 VARICP8014 VARICP8529 VARICP8730 VARICP7130 558-0004 558-0005 558-0007 558-0016 VARICP7454 558-0021 VARICP7620 558-0008 558-0026 VARICP7629 VARICP7635 VARICP7645 VARICP7646 VARICP7647 VARICP7649 VARICP7657 568-0714 VARICP7666 VARICP7670 VARICP7671 558-0010 VARICP7676 VARICP7685 VARICP7688 VARICP7693 VARICP7695 VARICP7700 VARICP7709 VARICP7710 558-0011 568-0716 558-0013 558-0023 VARICP7730 558-0015 VARICP7739 558-0024 VARICP7750 VARICP7758 VARICP7760 VARICP7762 VARICP7770 VARICP7829 VARICP7895 558-0018 558-0019 VARICP7917 VARICP7926 VARICP7936 VARICP7937 VARICP7946 VARICP7947 VARICP7977 VARICP8510 VARICP8521 VARICP8530 VARICP8540 VARICP8541 VARICP8560 VARICP8562 VARICP8673 VARICP8674 VARICP8677 VARICP8689 VARICP8690 VARICP8710 VARICP8712 VARICP8716 VARICP8722 VARICP8735 VARICP8736 VARICP8737 VARICP8741 VARICP8742 VARICP8743 VARICP8744 VARICP8751 VARICP8752 VARICP8753 VARICP8754 VARICP8756 VARICP8760 VARICP8761 VARICP8762 VARICP8770 VARICP8771 VARICP8772 VARICP8775 VARICP8780 VARICP8781 VARICP8791 VARICP8796 VARICP8799 VARICP8842 VARICP8870 VARICP8871 VARICP7681 VARICP7761 VARICP8431 VARICP7702 VARICP7680 558-0012 558-0014 558-0025 VARICP7135 VARICP7140 VARICP6666 VARICP7196 VARICP7630 VARICP7442 VARICP7443I5 VARICP7453I5 VARICP7732 VARICP7740 VARICP7749 VARICP7749I5 VARICP7750I5 VARICP7751I5 VARICP7741I5 VARICP7719 VARICP7720 VARICP7789 VARICP7809 VARICP7779 VARICP7697 VARICP7769 VARICP7770I5 VARICP7771 VARICP7772 VARICP7631 VARICP7621 VARICP7622 VARICP7636 VARICP7637 VARICP7608 VARICP8430 VARICP7680I5 VARICP7682 VARICP7684 VARICP8542 VARICP7690 VARICP7690I5 VARICP7691I5 VARICP7692 VARICP7694 VARICP7696 VARICP7640 VARICP7641 VARICP8511 VARICP8531 VARICP7839 VARICP7813 VARICP7896 VARICP7884 VARICP8690I5 VARICP8676 VARICP8678 VARICP8685 VARICP8687 VARICP8687I5 VARICP8688I5 VARICP8675 VARICP8735I5 VARICP8736I5 VARICP8741I5 VARICP8750 VARICP7656 VARICP7667 VARICP7670I5 VARICP7672 VARICP6680 VARICP6670 VARICP7150 VARICP6681 VARICP6671 VARICP7340 VARICP7160 VARICP7170 VARICP7310 VARICP7311 VARICP7685I5 VARICP8774
Capillary columns, CP-Sil
Columnas de cromatografía
The CP-Si™l line consists of a range of stationary phases which are based on a polysiloxane backbone. Functional groups are added to the polymer to introduce selectivity. Each polymer used for the CP-Sil line is manufactured in-house and is optimised to obtain highest degree of reproducibility.