Preparative columns, PLgel™
Proveedor: VARIAN
PL1210-6120 PL1210-6140 PL1210-1120 PL1210-6100 PL1210-6104 PL1210-6115 PL1210-6125 PL1210-6130 PL1210-6150 PL1210-6160
VARIPL1210-6120EA 6660 EUR
VARIPL1210-6120 VARIPL1210-6140 VARIPL1210-1120 VARIPL1210-6100 VARIPL1210-6104 VARIPL1210-6115 VARIPL1210-6125 VARIPL1210-6130 VARIPL1210-6150 VARIPL1210-6160
Preparative columns, PLgel™
Columnas de cromatografía
These preparative columns is ideal to fractionate polymers, isolate components in a polymer formulation or simplify mixtures of relatively small molecules in complex matrices.
  • Excellent column efficiency provides optimum resolution
  • High loading can isolate milli grams amounts for further study
  • Over 10 times scale up permits efficient quantification
It is packed with the same rigid, high performance media as the analytical columns so that mixtures of materials are easily separated on the basis of size, preferably in a low boiling organic solvent. TheyMore Product Information