Ion exchange chromatography columns, PL-SAX and PL-SCX
Proveedor: VARIAN
PL1745-3102 PL1745-3103 PL1745-3702 PL1745-3703 PL1751-3102 PL1751-3103 PL1751-3702 PL1751-3703 PL1845-2102 PL1845-2103 PL1845-3102 PL1845-3103 PL1851-2102 PL1851-2103 PL1851-3102 PL1851-3103 PL1245-1102 PL1245-1103 PL1245-3102 PL1245-3103 PL1245-3702 PL1245-3703 PL1251-1102 PL1251-1103 PL1251-3102 PL1251-3103 PL1251-3702 PL1251-3703
VARIPL1745-3102EA 20420 EUR
VARIPL1745-3102 VARIPL1745-3103 VARIPL1745-3702 VARIPL1745-3703 VARIPL1751-3102 VARIPL1751-3103 VARIPL1751-3702 VARIPL1751-3703 VARIPL1845-2102 VARIPL1845-2103 VARIPL1845-3102 VARIPL1845-3103 VARIPL1851-2102 VARIPL1851-2103 VARIPL1851-3102 VARIPL1851-3103 VARIPL1245-1102 VARIPL1245-1103 VARIPL1245-3102 VARIPL1245-3103 VARIPL1245-3702 VARIPL1245-3703 VARIPL1251-1102 VARIPL1251-1103 VARIPL1251-3102 VARIPL1251-3103 VARIPL1251-3702 VARIPL1251-3703
Ion exchange chromatography columns, PL-SAX and PL-SCX
Columnas de cromatografía
These rigid, strong ion exchange materials are extremely hydrophilic that is ideal for purification of biomolecules. The PL-SAX and PL-SCX materials are totally polymeric and are chemically and thermally stable over a full range of HPLC conditions.
  • Ion exchange purifications over a wider pH range extend applications
  • HPLC flow rates and rapid equilibration reduce purification cycle times
  • Large pore size for improved mass transfer delivers high speed, high resolution purifications
  • The 1000 Å pore size is for high capacity purifications
  • The 4000 Å with