Digital Ceramic Magnetic Hotplate Stirrers, AREC 7
Proveedor: VELP SCIENTIFIC
F20500681 SA20500681 SB20500681 SC20500681
VELPF20500681EA 0 EUR
VELPF20500681 VELPSA20500681 VELPSB20500681 VELPSC20500681
Digital Ceramic Magnetic Hotplate Stirrers, AREC 7
Imánes de agitación Agitadores magnéticos con placas calefactoras
Powerful digital hotplate stirrer, equipped with a 177,8 mm square ceramic plate, designed to deliver precise temperature control, advanced stirring, improved safety features, and ease of use.
  • Advanced heating plate technology ensures temperature homogeneity and stability up to 550 °C
  • Precise thermoregulation of the medium with PT100/PT1000 probe (±1 °C) and VTF Digital Thermoregulator (±0,5 °C)
  • Thermoregulation ranges from room temperature to 350 °C with external PT100 and PT1000 probes
  • Brushless motor ensuring power and resistance over time
  • Stirs up to 15