Stands for Imhoff sedimentation cones
Proveedor: VITLAB
81056
VITL81056EA 143 EUR
VITL81056
Stands for Imhoff sedimentation cones
Soportes de retortas
Sedimentation racks, made of PMMA. Holds two Imhoff sedimentation cones. Base plate with depression for exact vertical positioning of the sedimentation cone.