Water purification system, arium® advance EDI
Proveedor: Sartorius
H2O-EDI-2-T H2O-EDI-2-B H2O-EDI-1-T H2O-EDI-1-B
171-1296EA 8820 EUR
171-1296 VIVAH2O-EDI-2-B 171-1297 171-1298
Water purification system, arium® advance EDI
Purificación de agua Sistemas de agua pura y ultra pura
The arium® advance EDI provides Type 2 water in consistently high quality. The innovative iJust makes the best use of environmental resources by automatically optimizing water consumption. Unlike conventional water purification systems, the unique display with touch functions in combination with the intuitive menu navigation ensures easy operation.
  • Applied for microbiological media and reagents
  • Solutions for chemical analysis and synthesis, histology, ELISA, RIA and buffer solutions
  • Feed water for laboratory devices like autoclaves and dishwashers
  • Premium