VWR® Media Bottles, Square, PETG
Proveedor: VWR Collection
VWRI210-000057EA 177 EUR
VWRI210-000057 VWRI210-000071 VWRI210-000070 VWRI210-000068 VWRI210-000056 VWRI210-000082 VWRI210-000066 VWRI210-000063 VWRI210-000073 VWRI210-000084 VWRI210-000064 VWRI210-000081 VWRI210-000069 VWRI210-000058 VWRI210-000080 VWRI210-000062 VWRI210-000065 VWRI210-000075 VWRI210-000059 VWRI210-000060 VWRI210-000074 VWRI210-000067 VWRI210-000079 VWRI210-000072 VWRI210-000076 VWRI210-000083 VWRI210-000061 VWRI210-000078 VWRI210-000077 VWRI210-000055
VWR® Media Bottles, Square, PETG
Frascos Frascos de medios
Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and autoclaving steps with sterile and non-pyrogenic.
  • Made from 100% virgin PETG material, durable and break resistant
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability
  • Arranged in shrink-wrapped trays and polybag for