VWR® Centrifuge Tube Rack, 15 and 50 ml
Proveedor: VWR Collection
VWRI479-2289EA 693 EUR
VWRI479-2289
VWR® Centrifuge Tube Rack, 15 and 50 ml
Gradillas Tube Racks
Racks are specially designed to provide an ideal storage solution for large quantities of samples in 50 ml centrifuge tubes.
  • Easy access and storage
  • Security lock devices are available in each rack
  • Made of corrosion resistant pharmaceutical grade stainless steel
Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer.