Depyrogenated Dilution Tubes
Proveedor: FUJIFILM WAKO CHEMICALS
DL-13100
WAKFDL-13100EA 0 EUR
WAKFDL-13100
Depyrogenated Dilution Tubes
Tampones Tampones biológicos
These depyrogenated dilution tubes are made of high-quality borosilicate glass depyrogenated at 250 °C.
  • Endotoxin-free test tubes (<0,001 EU/tube)
  • Designed for all testing methods
  • Made with borosilicate glass
  • Can be used for general lab testing
This results in endotoxin-free tubes that can perform successful LAL dilution series, from creating the standard curve to sample dilutions, with minimal chance of potential contamination.