Tarjeta de recolección de muestras desmontable Whatman™ 903 Proteinsaver
Proveedor: Whatman products (Cytiva)
10534320
WHAT10534320EA 216 EUR
WHAT10534320
Tarjeta de recolección de muestras desmontable Whatman™ 903 Proteinsaver
Sistemas para diagnóstico Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
903 sample collection card with four sample spots.
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with four half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75 - 80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining coverMore Product Information