Extraction thimbles, high purity, glass, Whatman™
Proveedor: Whatman products (Cytiva)
2814-199 2814-432 2814-300 2814-259 2814-533
WHAT2814-199EA 295 EUR
WHAT2814-199 515-9012 WHAT2814-300 WHAT2814-259 WHAT2814-533
Extraction thimbles, high purity, glass, Whatman™
Aparatos de extracción Dedales de extracción
Cytiva's Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
  • Resistant to temperatures up to 500 °C
  • Binder-free, no chemical additives
  • Designed to fit most commercially available Soxhlet extractors
  • Effective when used with solvents incompatible with cellulose materials
Glass Soxhlet extraction thimble applications:
Analysis of hot and acidic gases during emission control and smoke stack gasMore Product Information