Arsine generator apparatus, WHEATON®
Proveedor: DWK Life Sciences
180034 180030 970440
WHEA180034EA 145 EUR
WHEA180034 WHEA180030 WHEA970440
Arsine generator apparatus, WHEATON®
Digestores Aparato de detección de trazas de elementos
Designed for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method.
  • Assembled apparatus is freestanding and easy to use
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.