Erlenmeyer flask with screw cap, WHEATON®
Proveedor: DWK Life Sciences
635134
WHEA635134EA 47.3 EUR
WHEA635134
Erlenmeyer flask with screw cap, WHEATON®
Matraces Matraces Erlenmeyer
These flasks, made of low extractable borosilicate glass, are used for continuous extraction of analytes from a solid into an organic solvent.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes