Bomb sampler, Wheaton®
Proveedor: DWK Life Sciences
885200
WHEA885200EA 2480 EUR
WHEA885200
Bomb sampler, Wheaton®
Muestreadores Muestreadores de líquidos
An efficient and unique composite sampling device made of stainless steel.
  • Cylindrical chamber
  • Weighted plunger that seals the chamber
  • Cable attachments for suspending the apparatus and activating the sampling device
Suitable for collecting liquid from top, bottom or any intermediate level of storage tanks, tank cars, lagoons, ponds and wells.
Certificaciones: EPA composite sampling methods 'Test Methods for Evaluating Solid Waste 1.1.4.1'.