NMR tubes, low pressure/vacuum, SP Wilmad-LabGlass
Proveedor: SP Industries
513-7LPV-7 307-LPV-7 507-LPV-7 507-LPV-8 528-LPV-7
WILM513-7LPV-7EA 213 EUR
WILM513-7LPV-7 WILM307-LPV-7 WILM507-LPV-7 WILM507-LPV-8 WILM528-LPV-7
NMR tubes, low pressure/vacuum, SP Wilmad-LabGlass
Tubos Tubos NMR
Borosilicate glass
  • 4X larger sealing surface increases lifetime and reliability
  • Greaseless PTFE piston provides a 100% contamination-free seal
  • Axial symmetric design guarantees its application in spinning experiments
  • More economical to reduce experimental costs
These tubes are ideal for anaerobic and gas-tight NMR and EPR experiments. They offer a convenient flame-free sealing solution for air sensitive or volatile liquid samples. The robust sealing system allows pressure build-up inside sample.
Certificaciones: Glass conforms to USP Type I andMore Product Information