Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Proveedor: SP Industries
LG-6655-100
WILMLG-6655-100EA 784 EUR
WILMLG-6655-100
Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Componentes para destilación Sets de destilación
This distilling apparatus is used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water.
  • The Clevenger type receiver trap included in this apparatus is for use when the oils are lighter than water
  • The condenser rests on top of the receiver
  • There is a discharge stopcock below the return tube
  • Flask and receiver are connected by a standard taper 24/40 joint
The receiver, graduated to 5 ml with 0,1 ml increments, has a tube connecting the lower part below the scale with the top of the vapour tube to returnMore Product Information