Pouring rings for laboratory bottles
Proveedor: witeg Labortechnik
0.175.201
WITG0.175.201EA 0.4 EUR
WITG0.175.201
Pouring rings for laboratory bottles
Tapas Tapones de botellas
Pouring rings, made of polypropylene.
  • For DIN GL threads
  • Heat resistant up to +140 °C
  • Good chemical resistance