Flasks, iodine, with collar, with hollow glass stopper
Proveedor: witeg Labortechnik
2.438.500
WITG2.438.500EA 76 EUR
WITG2.438.500
Flasks, iodine, with collar, with hollow glass stopper
Matraces Frascos para yodo
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area. Hollow glass stopper with extra long stem.