Safety wash bottles, narrow neck, with printed name
Proveedor: witeg Labortechnik
7.169.109 7.169.110 7.169.111 7.169.113 7.169.114 7.169.116 7.169.142 7.169.143
WITG7.169.109EA 5 EUR
WITG7.169.109 WITG7.169.110 WITG7.169.111 WITG7.169.113 WITG7.169.114 WITG7.169.116 WITG7.169.142 WITG7.169.143
Safety wash bottles, narrow neck, with printed name
Frascos Frascos de lavado
LDPE, yellow, with coloured screw caps.
  • With hazard symbols and descriptions of hazards as required by the hazardous substance ordinance
  • Good chemical resistance
  • Good thermal resistance
With safety delivery jet.