Reciprocating shaker, SHR digital
Proveedor: witeg Labortechnik
DH.WSR04022
WITGDH.WSR04022EA 1030 EUR
WITGDH.WSR04022
Reciprocating shaker, SHR digital
Agitadores de laboratorio Agitadores Recíprocos
These digital reciprocating shakers with programmable digital feedback control system are ideal for culturing cells in hybridisation, staining and destaining gels. Also used in combinatorial chemistry.
  • Brushless DC motor (low noise and long life cycle)
  • High precision digital rotation speed control
  • Over-Load protection, storage function and alarm
  • Programmable digital feedback controller with Jog-Shuttle switch (turn + push)
The optimal platform rotation helps for intensive mixing with convenient setting of timer and speed using jog shuttle switch. ForMore Product Information