Ion selective electrode system, AmmoLyt® Plus
Proveedor: WTW ONLINE
107042
WTWO107042EA 900 EUR
WTWO107042
Ion selective electrode system, AmmoLyt® Plus
Electrodos Electrodos selectivos de iones (ISE)
This system – consisting of a robust, digital armature with ammonium replacement electrode and ammonium reference electrode – allows ammonium measurement directly in the medium, without sample preparation. The system is designed for connection to the IQ SENSOR NET multiparameter measuring system.
  • Calibration-free, long term stable
  • With integrated temperature sensor
  • Short response time
  • With screw / plug connector for connection of the SACIQ sensor connection cable
Información para pedidos: Supplied with unequipped sensor AmmoLyt® Plus 700 IQ,More Product Information