MEM non essential amino acids 100X
Proveedor: Biowest
X0557-100
X0557-100EA 17.5 EUR
X0557-100
MEM non essential amino acids 100X
Suplementos y aditivos de cultivo celular
Concentrated solutions of amino acids, vitamins or non essential amino acids can be added to a basal medium to enrich it and the resulting medium used as a classical MEM or BME. The final concentration of the MEM or BME must be 1X.