LED illumination systems, HB 100, for microscopes
Proveedor: ZEISS
4230119901000
ZEIS4230119901000EA 0 EUR
ZEIS4230119901000
LED illumination systems, HB 100, for microscopes
Luces Iluminación LED
HBO 100 illuminator, self-adjusting with lamp mount and collector, including heat-reflecting filter and 2 m connection cable. Automatic lamp alignment.
  • Automatic lamp alignment