Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, I/P 26; 25 ft
Proveedor: Avantor Fluid Handling
Reduce the risk of particulate entrapment with smooth inner surface.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory Approvals to match your unique application
- No leachable additives protects sensitive and costly fluids
- I/P® 26 tubing has a 1/4” (6.4 mm) inner diameter
- Accepts 1/4” hose barb for tubing connection
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications<br />Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
See 96119-82 for the irradiated version of this tubing.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.