Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, I/P 73; 25 ft
Proveedor: Avantor Fluid Handling
VWR Item#MFLX96410-73
Your Organization's Price:
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • No leachable additives protects sensitive and costly fluids
  • I/P® 73 tubing has a 3/8” (9.5 mm) inner diameter
  • Accepts 3/8” hose barb for tubing connection
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications<br />Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
See 96119-73 for the irradiated version of this tubing.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.