Masterflex® Sanitary Clamp with White Knob, Nylon, 1/2" to 3/4"; 1/EA
Proveedor: Avantor Fluid Handling
Compatible with metal and plastic Tri-Clamp® fittings.
- Excellent chemical resistance; low leachable characteristics
- Autoclavable
- Accepts 1/2" to 3/4" tubing
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.