Masterflex® Sanitary Clamp with White Knob, Nylon, 2"; 1/EA
Proveedor: Avantor Fluid Handling
VWR Item#MFLX30508-25
Your Organization's Price:
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance; low leachable characteristics
  • Autoclavable
  • Accepts up to 2" tubing
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.