Masterflex® I/P® Spooled Precision Pump Tubing, BioPharm Platinum-Cured Silicone, I/P 73; 100 ft
Proveedor: Avantor Fluid Handling
Smooth inner surface designed to reduce risk of particle entrapment and contamination.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- I/P® 73 tubing has a 3/8” (9.5 mm) inner diameter
- Accepts 3/8” hose barb for tubing connection
Choose our BioPharm Silicone platinum-cured tubing for purity, very low extractables, and an ultra-smooth inner surface. Ideal for pharmaceutical and biotechnological applications, this tubing complies with USP class VI and 3A sanitary standards.<br /><br />The ultra-smooth inner surface minimizes particle entrapment and microscopic build-up during fluid transfer. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.<br /><br />Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.
I/P® Precision pump tubing is compatible with Masterflex® Standard and Easy-Load pump heads.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.