Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
Proveedor: Avantor Fluid Handling
VWR Item#MFLX30620-24
Your Organization's Price:
Ensure a tight seal every time.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 3/8" ID; 1/EA
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. <br />Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.