Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kit for Precision Tubing
Proveedor: Avantor Fluid Handling
Enhance the life of your pump head with the replacement retainer kits.
- Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
- Suitable for use with Easy-Load® II pump heads 77200-50, -60, and 77201-60
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
<b>Información de suministro: </b>Includes two small upper retainers, two small lower retainers, and two compression springs.