Masterflex® Transfer Tubing, Polytetrafluoroethylene, 3/8" ID x 1/2" OD; 100 ft
Proveedor: Avantor Fluid Handling
VWR Item#MFLX95231-14
Your Organization's Price:
Available in spooled rolls for increased versatility.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 3/8" ID x 1/2" OD
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.