Masterflex® L/S® High-Performance Precision Pump Tubing, Puri-Flex®, L/S 24; 25 ft
Proveedor: Avantor Fluid Handling
Long life tubing for any Masterflex® pump used in your pharma and bioprocess applications.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 24
- Certificate of compliance included with each individual unit upon shipment
<B>Applications: </B>Cell and tissue culture work, pharmaceutical biotech, general-purpose lab uses requiring high performance and long life, high-purity applications requiring no contact with animal-derived components<br /><B>Characteristics: </B>Heat weldable, exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, translucent<br /><B>Certifications: </B>FDA compliant, meets USP Class VI requirements, manufactured according to GMP<br /><B>Temperature range: </B>–58 to 275 °F (–50 to 135 °C)<br /><B>Sterilize: </B>Autoclave, ethylene oxide, or gamma irradiation
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.