Masterflex® Ball valve, 3-way, 3/8" barb - PVC w/Viton® seals
Proveedor: Avantor Fluid Handling
VWR Item#MFLX01377-74
Your Organization's Price:
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.