Masterflex® Ball Valves, Three-way, PVC, 1/2" ID Hose Barb
Proveedor: Avantor Fluid Handling
VWR Item#MFLX01379-07
Your Organization's Price:
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.