Rotary drum pump, High Volume 17 GPM, Cast-Iron
Proveedor: Avantor Fluid Handling
Dispense a variety of liquids compatible with polypropylene, Ryton® PPS, cast iron, aluminum, stainless steel, or PTFE.
- Lightweight and easy to use
- Smooth, non-pulsating flow
- Operate either in forward or reverse direction with a discharge hose
Polypropylene rotary pump (06510-00) is excellent for most organic acids, waxes, plating solutions, detergents and water soluble inorganic chemicals. Ryton PPS rotary pump (06510-05) is able to dispense aggressive chemicals such as esters, ketones and hydrocarbons. Ryton PPS pumps include a stainless steel discharge tube with a 3-piece stainless steel suction tube. Stainless steel pump (06510-10) with PTFE seal is suitable for pumping chlorinated or petroleum-based solvents. For larger flows with compatible liquids, there are now cast iron (06510-17) and aluminum (06510-19) pumps.<br /><br />Call our Application Specialists to order optional 1" PVC 5-ft tubing with hose clamps.
<b>Información de suministro: </b>Adapter type 11/2" x 2" NPT bung.