Hand-Operated Piston Dispensing Drum Pump, Polypropylene, 16 to 64 Strokes/Gal
Proveedor: Avantor Fluid Handling
VWR Item#MFLX06512-00
Your Organization's Price:
Economical piston pumps are ideal for a variety of fluids.
  • Adaptable to most pails and drums
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.