GF Signet 161.514.614 Type 514 PVC Manual Diaphragm Valve, 1", 32.5 Cv
Proveedor: Avantor Fluid Handling
Optimized valve body provides double the C<sub>V</sub> values.
- Two-color positition indicator on handwheel
- Elimination of bolts removes corrosive weak points
- Mount vertically or horizontally
- PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150 °F (65 °C) and feature true union socket connections.