GF Signet 161.514.617 Type 514 Manual Diaphragm Valve, PVC, 2" True Union Socket
Proveedor: Avantor Fluid Handling
VWR Item#MFLX07390-56
Your Organization's Price:
Optimized valve body provides double the C<sub>V</sub> values.
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150 °F (65 °C) and feature true union socket connections.