Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Red, 1/16" ID x 1/8" OD; 12 ft
Proveedor: Avantor Fluid Handling
Quickly identify lines.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Tubing size: 1/16" ID x 1/8" OD
- Color-coded red for quick identification
Applications: Ideal for dedicated or multiple lines—provides quick and easy line identification.<br />Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque.<br />Certifications: None.<br />Temperature range: -400 to 500 °F (-240 to 260 °C).<br />Sterilize: Autoclave, ethylene oxide, or dry heat.