Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Red, 3/16" ID x 1/4" OD; 12 ft
Proveedor: Avantor Fluid Handling
VWR Item#MFLX96130-04
Your Organization's Price:
Quickly identify lines.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 3/16" ID x 1/4" OD
  • Color-coded red for quick identification
Applications: Ideal for dedicated or multiple lines—provides quick and easy line identification.<br />Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque.<br />Certifications: None.<br />Temperature range: -400 to 500 °F (-240 to 260 °C).<br />Sterilize: Autoclave, ethylene oxide, or dry heat.