CPC® Miniature Quick-Disconnect Fitting, Panel-Mount Hose Barb Body, Polycarbonate, Straight-Through, 3 mm ID, SMFPMM3; 50/PK
Proveedor: Avantor Fluid Handling
VWR Item#MFLX06363-97BLK
Your Organization's Price:
No more kinked tubing - allows tubing to rotate freely.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle ¹/₁₆" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.<br /><br />For a complete coupling, order one fitting body and one insert.