[EN]FITTING SANITARY TCXB 3/4X1/2 SS EA 1 * 1 UN
Proveedor: Avantor Fluid Handling
VWR Item#MFLX30510-19
Your Organization's Price:
High-quality tri-clamp to hose barb fittings in 316 stainless steel for sanitary applications.
  • Constructed from 316 stainless steel for excellent corrosion resistance and durability
  • Designed to meet sanitary standards, ensuring hygienic fluid transfer in critical applications
  • Secure tri-clamp design allows for reliable attachment to various equipment and systems
  • Tailored hose barb shape and size options to accommodate a wide range of hose sizes
  • Precision engineered and designed for use with tubing clamps to ensure tight seals for leak-free connections, maintaining product integrity
  • Suitable for various industries including food and beverages, pharmaceuticals, and more
  • Capable of handling demanding operating conditions with resistance to high temperatures and pressures
  • Designed to work seamlessly with existing equipment, enhancing overall system compatibility
Elevate your fluid transfer systems with these premier sanitary tri-clamp to hose barb fittings in 316 stainless steel. Engineered for excellence, these fittings combine top-grade materials with hygienic design, ensuring unparalleled durability, corrosion resistance, and leak-free performance. Their versatile compatibility with various hose types and sizes, seamless tri-clamp attachment, and easy maintenance make installation a breeze, and supporting industries ranging from food and beverage to pharmaceuticals. Trust in these fittings to enhance operational efficiency, promote system integrity, and set new benchmarks for fluid management excellence.