Cleanroom anti-fatigue drainage mats, Ergomat® Endurance
Proveedor: ERGOMAT
EX0220 EX0209 EX0206 EX0306 EX0320 EX0304 EX0412 EX0406 EX0303 EX0309 EX0312 EX0204 EX0212 EX0404 EX0409
115-0920EA 1190 EUR
115-0920 115-0919 115-0918 115-0922 115-0923 115-0921 115-0925 115-0924 ERGOEX0303 ERGOEX0309 ERGOEX0312 ERGOEX0204 ERGOEX0212 ERGOEX0404 ERGOEX0409
Cleanroom anti-fatigue drainage mats, Ergomat® Endurance
Alfombrillas Alfombrillas antifatiga
The Ergomat Endurance is a pure nitrile rubber drainage mat designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave resulting from employee short term disability. This mat is ideal for very wet environments.
  • Excellent anti-slip and drainage properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevents curling, eliminates trip hazards
  • Raw virgin material guarantees durability and ergonomic benefits
  • Free from silicone and latex
Suitable for use in ISO 5, Class 100 cleanrooms.
Colour: Black