Ion exchange HPLC columns, Bio
Proveedor: Agilent
5190-2439 5190-2421 5190-2423 5190-2424 5190-2425 5190-2426 5190-2427 5190-2431 5190-2435 5190-2440 5190-2443 5190-2444 5190-2451 5190-2452 5190-2453 5190-2454 5190-2459 5190-2460 5190-2461 5190-2432 5190-2465 5190-2463 5190-2494 5190-2467 5190-2471 5190-2472 5190-2473 5190-2479 5190-2480 5190-2485 5190-2487 5190-2491 5190-2493 5190-2495 5190-2499 5190-2500 5190-2464 5190-2492
HEWL5190-2439EA 2113 EUR
HEWL5190-2439 HEWL5190-2421 HEWL5190-2423 HEWL5190-2424 HEWL5190-2425 HEWL5190-2426 HEWL5190-2427 HEWL5190-2431 HEWL5190-2435 HEWL5190-2440 HEWL5190-2443 HEWL5190-2444 HEWL5190-2451 HEWL5190-2452 HEWL5190-2453 HEWL5190-2454 HEWL5190-2459 HEWL5190-2460 HEWL5190-2461 HEWL5190-2432 HEWL5190-2465 HEWL5190-2463 HEWL5190-2494 HEWL5190-2467 HEWL5190-2471 HEWL5190-2472 HEWL5190-2473 HEWL5190-2479 HEWL5190-2480 HEWL5190-2485 HEWL5190-2487 HEWL5190-2491 HEWL5190-2493 HEWL5190-2495 HEWL5190-2499 HEWL5190-2500 HEWL5190-2464 HEWL5190-2492
Ion exchange HPLC columns, Bio
Columnas de cromatografía
These HPLC columns are packed with polymeric, nonporous, ion exchange particles and are designed for high resolution, high recovery and highly efficient separations of peptides, oligonucleotides and proteins.
  • Uniform, densely packed ion exchange functional groups are chemically bonded to hydrophilic layer to increase column capacity
  • Particles, coating and bonding are resistant to high pressures, promoting higher resolution and faster separations
  • Multiple ion exchange groups are captured on one anchoring to increase capacity
It offers strong cationMore Product Information