SPE columns, CHROMABOND® NH₂
Proveedor: MACHEREY-NAGEL
730180 730180G 730626 730626G 730031 730033 730033G 732033
MANA730180EA 142 EUR
MANA730180 MANA730180G MANA730626 MANA730626G MANA730031 MANA730033 MANA730033G MANA732033
SPE columns, CHROMABOND® NH₂
Columnas de cromatografía
CHROMABOND® NH₂ is an aminopropyl modified silica phase for SPE.
  • Polar phase
  • Weak anion exchanger
  • Interaction with moderately polar and ionic compounds
Recommended for trace elements and lipids.
Order Now
ESPECIFICACIONES
Environmentally Preferable
Carga de carbono 3,5%
Rango de pH 2 - 8
Surface area 500 m²/g