Hexaammineruthenium trichloride (≥32,1% Ru)
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride
10511.03
10511.06
10511.01
10511.03EA
207
EUR
10511.03
10511.06
10511.01
Hexaammineruthenium trichloride (≥32,1% Ru)
Hexaammineruthenium trichloride
CAS No.: 14282-91-8
Molecular Formula: Ru(NH3)6Cl3
Formula Weight: 309.61
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale yellow
UN No.: UN3288
MDL No.: MFCD00011478
Formula:
H₁₈Cl₃N₆Ru MW: 309,61 g/mol |
Número MDL:
MFCD00011478 Núm. CAS: 14282-91-8 EINECS: 238-176-8 UN: 3288 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...