Fmoc-L-phenylalanine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

F0297-25G F0297-5G
TCIAF0297-25GEA 52 EUR
TCIAF0297-25G TCIAF0297-5G
Fmoc-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
Fmoc-L-phenylalanine
Formula: C₂₄H₂₁NO₄
MW: 387,4 g/mol
Número MDL: MFCD00037128
Núm. CAS: 35661-40-6
EINECS: 252-661-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0774c35fd592aaeb54f558b2766aec87ad65851d&vid=0774c35fd592aaeb54f558b2766aec87ad65851d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 187.0 deg-C
Specific rotation [a]20/D -36.0 to -40.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR