9-Phenylcarbazole ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
N-Phenylcarbazole
P1492-1G
P1492-25G
P1492-5G
TCIAP1492-1GEA
17
EUR
TCIAP1492-1G
TCIAP1492-25G
TCIAP1492-5G
9-Phenylcarbazole ≥98.0% (por GC)
9-Phenylcarbazole
Formula:
C₁₈H₁₃N MW: 243,30 g/mol |
Número MDL:
MFCD00004965 Núm. CAS: 1150-62-5 EINECS: 214-564-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...