Iminostilbene ≥98.0% (by GC, total nitrogen)

Proveedor: TCI
Atención

I0414-25G I0414-5G
TCIAI0414-25GEA 59 EUR
TCIAI0414-25G TCIAI0414-5G
Iminostilbene ≥98.0% (by GC, total nitrogen)
Iminostilbene
Formula: C₁₄H₁₁N
Número MDL: MFCD00005071
Núm. CAS: 256-96-2
EINECS: 205-970-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8bd049aace7d8b1d0193aac6c328a0319cb59b5&vid=f8bd049aace7d8b1d0193aac6c328a0319cb59b5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 197.0 to 201.0 deg-C
Solubility in Dioxane almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR