Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: BOP-Cl

B1213-25G B1213-5G
TCIAB1213-25GEA 106 EUR
TCIAB1213-25G TCIAB1213-5G
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0% (by titrimetric analysis)
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
Formula: C₆H₈ClN₂O₅P
MW: 254,6 g/mol
Número MDL: MFCD00010077
Núm. CAS: 68641-49-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48887a7481caf4fec003fc0e9f33fdd890f0bb8f&vid=48887a7481caf4fec003fc0e9f33fdd890f0bb8f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR